ENA402266923 Enamine

8-chloro-2-ethyl-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD25371718
  • Cas Number : 1461707-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131457

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131457
Availability details for
Quantity
  • Availability
  • Add to Cart