ENA402266926 Enamine

2-[(1-phenylethyl)amino]propanoic acid hydrochloride

  • MDL number MFCD25371722
  • Cas Number : 1485818-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131479
Availability details for
Quantity
  • Availability
  • Add to Cart