ENA402266927 Enamine

2-[6-(methylsulfanyl)hexyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD25371723
  • Cas Number : 59501-75-6
  • Molecular Weight : 277.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131483
Availability details for
Quantity
  • Availability
  • Add to Cart