ENA402266932 Enamine

2-[3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-2-methoxypropyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD25371729
  • Cas Number : 856832-79-6
  • Molecular Weight : 364.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131653
Availability details for
Quantity
  • Availability
  • Add to Cart