ENA402266937 Enamine

benzyl({1-[3,5-bis(trifluoromethyl)phenyl]-2,2,2-trifluoroethyl})amine

  • MDL number MFCD25371735
  • Cas Number : 1461708-64-4
  • Molecular Weight : 401.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131697
Availability details for
Quantity
  • Availability
  • Add to Cart