ENA402266938 Enamine

tert-butyl N-{1-[(3,5-dichlorophenyl)methyl]piperidin-4-yl}carbamate

  • MDL number MFCD25371736
  • Cas Number : 1461708-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131731
Availability details for
Quantity
  • Availability
  • Add to Cart