ENA402266940 Enamine

3-(3-methyl-1,2,4-oxadiazol-5-yl)morpholine hydrochloride

  • MDL number MFCD25371740
  • Cas Number : 1461708-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131817
Availability details for
Quantity
  • Availability
  • Add to Cart