ENA402266943 Enamine

benzyl N-(3-{6,8-dioxo-5,7-diazaspiro[3.4]octan-7-yl}propyl)carbamate

  • MDL number MFCD25371749
  • Cas Number : 1461707-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131912
Availability details for
Quantity
  • Availability
  • Add to Cart