ENA402266946 Enamine

2-(3-methoxyphenyl)-3-phenylpyrrolidine hydrochloride

  • MDL number MFCD25371753
  • Cas Number : 1461715-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131935

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131935
Availability details for
Quantity
  • Availability
  • Add to Cart