ENA402266949 Enamine

ethyl 2-[2-(4-bromo-2-ethylphenyl)hydrazin-1-ylidene]propanoate

  • MDL number MFCD25371760
  • Cas Number : 1461726-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132021
Availability details for
Quantity
  • Availability
  • Add to Cart