ENA402266953 Enamine

methyl({2,2,2-trifluoro-1-[3-(trifluoromethyl)phenyl]ethyl})amine hydrochloride

  • MDL number MFCD25371765
  • Cas Number : 1461708-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198...200
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132056
Availability details for
Quantity
  • Availability
  • Add to Cart