ENA402266954 Enamine

(1-benzyl-4-ethylpiperidin-4-yl)methanol

  • MDL number MFCD25371768
  • Cas Number : 1461707-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132087
Availability details for
Quantity
  • Availability
  • Add to Cart