ENA402266955 Enamine

5-(4-methyl-1,3-thiazol-5-yl)-1,3-oxazole

  • MDL number MFCD25371769
  • Cas Number : 1461708-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132089
Availability details for
Quantity
  • Availability
  • Add to Cart