ENA402266956 Enamine

4,4,4-trifluorobutan-2-amine hydrochloride

  • MDL number MFCD25371770
  • Cas Number : 93339-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-179
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132091
Availability details for
Quantity
  • Availability
  • Add to Cart