ENA402266964 Enamine

(1,3-dimethyl-1H-pyrazol-4-yl)methanamine dihydrochloride

  • MDL number MFCD25371778
  • Cas Number : 1461713-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132162

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-132162
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart