ENA402266970 Enamine

3-(chloromethyl)-5-cyclobutyl-4-methyl-4H-1,2,4-triazole hydrochloride

  • MDL number MFCD25371784
  • Cas Number : 1461704-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132173
Availability details for
Quantity
  • Availability
  • Add to Cart