ENA402266973 Enamine

(5-cyclobutyl-4-methyl-4H-1,2,4-triazol-3-yl)methanamine dihydrochloride

  • MDL number MFCD25371788
  • Cas Number : 1461713-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132314
Availability details for
Quantity
  • Availability
  • Add to Cart