ENA402266979 Enamine

2-amino-1-cyclohexylethan-1-one hydrochloride

  • MDL number MFCD25371796
  • Cas Number : 349495-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132418

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132418
Availability details for
Quantity
  • Availability
  • Add to Cart