ENA402266981 Enamine

2-[(4-aminophenyl)methyl]-1,3-thiazole-4-carboxylic acid dihydrochloride

  • MDL number MFCD25371798
  • Cas Number : 1461708-11-1
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132422
Availability details for
Quantity
  • Availability
  • Add to Cart