ENA402266984 Enamine

2-chloro-N-(2-oxobutyl)acetamide

  • MDL number MFCD01940343
  • Cas Number : 1461707-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132624

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132624
Availability details for
Quantity
  • Availability
  • Add to Cart