ENA402266985 Enamine

2-(3-nitro-1H-pyrazol-1-yl)ethanimidamide hydrochloride

  • MDL number MFCD25371805
  • Cas Number : 1461713-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132625
Availability details for
Quantity
  • Availability
  • Add to Cart