ENA402266990 Enamine

3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]propan-1-amine hydrochloride

  • MDL number MFCD25371812
  • Cas Number : 1461708-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132658
Availability details for
Quantity
  • Availability
  • Add to Cart