ENA402266991 Enamine

N-amino-2-(3-bromophenyl)ethanimidamide hydrochloride

  • MDL number MFCD25371813
  • Cas Number : 1461707-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132660

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132660
Availability details for
Quantity
  • Availability
  • Add to Cart