ENA402266993 Enamine

3-{[benzyl(methyl)amino]methyl}benzoic acid hydrochloride

  • MDL number MFCD25371816
  • Cas Number : 1461713-70-1
  • Molecular Weight : 291.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132674

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132674
Availability details for
Quantity
  • Availability
  • Add to Cart