ENA402266998 Enamine

6-bromo-3,4-dihydro-2H-1-benzopyran-8-carbaldehyde

  • MDL number MFCD25371822
  • Cas Number : 1461705-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132773
Availability details for
Quantity
  • Availability
  • Add to Cart