ENA402267005 Enamine

6-amino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide

  • MDL number MFCD25371831
  • Cas Number : 67544-41-6
  • Molecular Weight : 244.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 280-285
Iupac Name 6-amino-5,6,7,8-tetrahydronaphthalen-2-ol hydrobromide
Inchi Code 1S/C10H13NO.BrH/c11-9-3-1-8-6-10(12)4-2-7(8)5-9;/h2,4,6,9,12H,1,3,5,11H2;1H
Inchi Key CELLWXZDJHUUTK-UHFFFAOYSA-N

Description

Melting Point 280-285
Purity 95
Salt Data Br
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-132972

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-132972
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart