ENA402267008 Enamine

5-aminopyridine-2-carboxylic acid hydrochloride

  • MDL number MFCD04114190
  • Cas Number : 78273-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254...256
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114806

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114806
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart