ENA402267010 Enamine

6-chloro-5-ethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03424274
  • Cas Number : 20295-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130036
Availability details for
Quantity
  • Availability
  • Add to Cart