ENA402267017 Enamine

1-{[4-(trifluoromethoxy)phenyl]methyl}piperidin-4-ol

  • MDL number MFCD15485328
  • Cas Number : 681482-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130208

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130208
Availability details for
Quantity
  • Availability
  • Add to Cart