ENA402267023 Enamine

5,6,7,8-tetrahydroquinolin-3-amine

  • MDL number MFCD15527170
  • Cas Number : 151224-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-98
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130295
Availability details for
Quantity
  • Availability
  • Add to Cart