ENA402267024 Enamine

5-bromo-2-(pyridin-3-yl)pyrimidine

  • MDL number MFCD16618820
  • Cas Number : 1240594-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130300
Availability details for
Quantity
  • Availability
  • Add to Cart