ENA402267027 Enamine

7-fluoro-2,3,4,5-tetrahydro-1H-1-benzazepin-5-ol

  • MDL number MFCD20668446
  • Cas Number : 1379213-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130336
Availability details for
Quantity
  • Availability
  • Add to Cart