ENA402267029 Enamine

tert-butyl 5-chloropentanoate

  • MDL number MFCD21117443
  • Cas Number : 105566-71-0
  • Molecular Weight : 192.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130358
Availability details for
Quantity
  • Availability
  • Add to Cart