ENA402267031 Enamine

3,3-dimethyl-2-[(trimethylsilyl)oxy]butanenitrile

  • MDL number MFCD16786601
  • Cas Number : 88522-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130372
Availability details for
Quantity
  • Availability
  • Add to Cart