ENA402267040 Enamine

2-amino-2-(naphthalen-2-yl)acetonitrile

  • MDL number MFCD09732818
  • Cas Number : 252637-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130453

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130453
Availability details for
Quantity
  • Availability
  • Add to Cart