ENA402267049 Enamine

ethyl 3-(N-ethyl-2,2,2-trifluoroacetamido)propanoate

  • MDL number MFCD25371629
  • Cas Number : 1461708-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130476
Availability details for
Quantity
  • Availability
  • Add to Cart