ENA402267052 Enamine

3-(3-fluoro-4-methoxyphenyl)propan-1-ol

  • MDL number MFCD09927328
  • Cas Number : 848349-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130489
Availability details for
Quantity
  • Availability
  • Add to Cart