ENA402267053 Enamine

6-bromo-8-fluoro-2-(trifluoromethyl)-1,4-dihydroquinolin-4-one

  • MDL number MFCD12114744
  • Cas Number : 1155601-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130491
Availability details for
Quantity
  • Availability
  • Add to Cart