ENA402267059 Enamine

tert-butyl N-(1,3-dioxaindan-4-yl)carbamate

  • MDL number MFCD09038117
  • Cas Number : 111081-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130571
Availability details for
Quantity
  • Availability
  • Add to Cart