ENA402267061 Enamine

1-(4-methoxy-3-methylphenyl)ethan-1-one

  • MDL number MFCD01673307
  • Cas Number : 10024-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 24-26
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-118793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118793
Availability details for
Quantity
  • Availability
  • Add to Cart