ENA402267063 Enamine

1-(4-fluorophenyl)-5-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11531312
  • Cas Number : 924644-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130601

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130601
Availability details for
Quantity
  • Availability
  • Add to Cart