ENA402267069 Enamine

4-iodo-1-methyl-3-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD24115723
  • Cas Number : 1461708-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130641
Availability details for
Quantity
  • Availability
  • Add to Cart