ENA402267071 Enamine

6-(cyclopentyloxy)pyridine-3-carboxylic acid

  • MDL number MFCD09740844
  • Cas Number : 509101-14-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130643

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130643
Availability details for
Quantity
  • Availability
  • Add to Cart