ENA402267072 Enamine

N-methylmorpholine-2-carboxamide

  • MDL number MFCD11656587
  • Cas Number : 135072-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130644

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-130644
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart