ENA402267073 Enamine

2-[4-(2-methoxyethoxy)phenyl]acetonitrile

  • MDL number MFCD09929422
  • Cas Number : 352547-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130647

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
hcodes T,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-130647
pictograms GHS06
hstatements H301,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart