ENA402267078 Enamine

2-(3-methoxyphenyl)-1,3-dithiolane

  • MDL number MFCD01729662
  • Cas Number : 82436-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130662
Availability details for
Quantity
  • Availability
  • Add to Cart