ENA402267080 Enamine

4-(tert-butoxy)-3,5-difluoroaniline

  • MDL number MFCD12822044
  • Cas Number : 942615-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130664

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130664
Availability details for
Quantity
  • Availability
  • Add to Cart