ENA402267082 Enamine

N-(prop-2-en-1-yl)acetamide

  • MDL number MFCD00607985
  • Cas Number : 692-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-120100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-120100
Availability details for
Quantity
  • Availability
  • Add to Cart