ENA402267084 Enamine

5-bromo-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD22421054
  • Cas Number : 54367-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130677

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130677
Availability details for
Quantity
  • Availability
  • Add to Cart