ENA402267093 Enamine

N-(2,2-dimethoxyethyl)-N'-methylethanediamide

  • MDL number MFCD08351201
  • Cas Number : 58108-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130736
Availability details for
Quantity
  • Availability
  • Add to Cart